5-(Methylthio)-1,2,4-thiadiazol-3-amine
Catalog # APOSOR322242-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:147.22 g/mol
Storage Temperature:Ambient
CAS Number:60093-10-9
Formula:C₃H₅N₃S₂
MDL Number:MFCD00799288