6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride
Catalog # APOSOR322184-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.69 g/mol
Storage Temperature:Ambient
CAS Number:20232-39-7
Formula:C₁₁H₁₄ClNO₂
MDL Number:MFCD00143492