
1,1-Dimethylbiguanide hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1115-70-4
:
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582
:165.625 g/mol
Melting Pt:221 °C
Density:1.36 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together