5-(4-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:109925-13-5
:
Formula:C₁₂H₁₁ClN₂O₂
MDL Number:MFCD00215390
:250.69 g/mol
Storage Temperature:Ambient
Frequently Bought Together