Order Entry
Italy
ContactUsLinkComponent
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate 95%
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate 95%
Catalog # APOSOR32174-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-1-oxo-2-(3-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylate 95%
Catalog # APOSOR32174-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.31 g/mol
Storage Temperature:Ambient
CAS Number:446276-03-5
Formula:C₁₇H₁₄N₂O₃
MDL Number:MFCD00140334