7-Chloro-1H-pyrrolo[3,2-b]pyridine 97%
Catalog # APOSOR321484-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:152.5800 g/mol
Storage Temperature:Ambient
CAS Number:357263-48-0
Formula:C₇H₅ClN₂
MDL Number:MFCD08234945