Order Entry
Italy
ContactUsLinkComponent
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%
Irritant
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%
Catalog # APOSOR321375-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 97%
Catalog # APOSOR321375-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.2000 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:800401-64-3
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD09878684