Order Entry
Italy
ContactUsLinkComponent
 
5-Bromo-2-hydroxy-3-picoline
Catalog # APOSOR3202-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2-hydroxy-3-picoline
Catalog # APOSOR3202-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one
 :
 :188.02 g/mol
Melting Pt:170…174 °C
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:89488-30-2
Formula:C₆H₆BrNO
MDL Number:MFCD03427657