Order Entry
Italy
ContactUsLinkComponent
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSOR318107-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
Catalog # APOSOR318107-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.16 g/mol
Storage Temperature:Ambient
CAS Number:916423-54-6
Formula:C₁₂H₁₂BrN₃
MDL Number:MFCD09258796