4-Chloro-6,7-dihydro-2-(methylthio)-7-phenyl-(5H)-pyrrolo[2,3-d]pyrimidine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:388572-68-7
:
Formula:C₁₃H₁₂ClN₃S
MDL Number:MFCD00202984
:277.78 g/mol
Storage Temperature:Ambiente
Frequently Bought Together