4-Chloro-7-cyclohexyl-6,7-dihydro-2-(methylthio)-(5H)-pyrrolo[2,3-d]pyrimidine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:388572-67-6
:
Formula:C₁₃H₁₈ClN₃S
MDL Number:MFCD00202983
:283.83 g/mol
Storage Temperature:Ambient
Frequently Bought Together