Order Entry
Italy
ContactUsLinkComponent
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR317001-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR317001-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.4700 g/mol
Storage Temperature:Ambient
CAS Number:1060815-90-8
Formula:C₆H₃ClIN₃
MDL Number:MFCD13189363