Order Entry
Italy
ContactUsLinkComponent
6-Bromo-1H-pyrrolo[3,2-c]pyridine 99%
Irritant
6-Bromo-1H-pyrrolo[3,2-c]pyridine 99%
Catalog # APOSOR31656-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-1H-pyrrolo[3,2-c]pyridine 99%
Catalog # APOSOR31656-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :197.0320 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1000342-71-1
Formula:C₇H₅BrN₂
MDL Number:MFCD08690134