
(4-Dimethylaminophenyl)di-tert-butylphosphine 97%
Catalog # APOSOR315484-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:265.38 g/mol
Melting Pt:57…61 °C
:GHS07
Storage Temperature:Ambient
CAS Number:932710-63-9
Formula:C₁₆H₂₈NP
MDL Number:MFCD09265102