Order Entry
Italy
ContactUsLinkComponent
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
Catalog # APOSOR315435-250MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl-4-oxo-3,9-diazaspiro[4.5]decane-9-carboxylate 98%
Catalog # APOSOR315435-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :254.33 g/mol
Storage Temperature:Ambient
CAS Number:923009-50-1
Formula:C₁₃H₂₂N₂O₃
MDL Number:MFCD11227060