Order Entry
Italy
ContactUsLinkComponent
2,6-Diethylphenyl isothiocyanate
2,6-Diethylphenyl isothiocyanate
Catalog # APOSOR315247-5G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Diethylphenyl isothiocyanate
Catalog # APOSOR315247-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :191.3 g/mol
Storage Temperature:Ambient
CAS Number:25343-69-5
Formula:C₁₁H₁₃NS
MDL Number:MFCD00041069