2,9-Diazaspiro[5.5]undecane-2-carboxylic acid tert-butyl ester
Catalog # APOSOR312332-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:254.38 g/mol
Storage Temperature:Ambient
CAS Number:189333-03-7
Formula:C₁₄H₂₆N₂O₂
MDL Number:MFCD11223510