2-Chloro-5,6,7,8-tetrahydro-1,6-naphthyridine hydrochloride 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:766545-20-4
:
Formula:C₈H₁₀Cl₂N₂
MDL Number:MFCD09835503
:205.0844 g/mol
Storage Temperature:Ambient
Frequently Bought Together