Order Entry
Italy
ContactUsLinkComponent
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride
Catalog # APOSOR310752-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Piperidin-3-yl-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine dihydrochloride
Catalog # APOSOR310752-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.24 g/mol
Storage Temperature:Ambient
CAS Number:1443423-29-7
Formula:C₁₂H₂₂Cl₂N₄
MDL Number:MFCD25975127