1'-Ethyl-6'-methyl-1'H-spiro[piperidine-4,2'-quinazolin]-4'(3'H)-one
Catalog # APOSOR310672-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:259.35 g/mol
Storage Temperature:Ambient
CAS Number:1351398-03-2
Formula:C₁₅H₂₁N₃O
MDL Number:MFCD20528963