Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Furyl)-1,3-butanedione
1-(2-Furyl)-1,3-butanedione
1-(2-Furyl)-1,3-butanedione
APOSOR310646-5G
:
Apollo Scientific
:
1-(2-Furyl)-1,3-butanedione
APOSOR310646-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
152.149 g/mol
:
Ambient
:
25790-35-6
:
C₈H₈O₃
:
MFCD00041759