
(S)-Methyl morpholine-3-carboxylate hydrochloride
Catalog # APOSOR308167-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.6174 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1447972-26-0
Formula:C₆H₁₂ClNO₃
MDL Number:MFCD09910333