Order Entry
Italy
ContactUsLinkComponent
tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate
Irritant
tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate
Catalog # APOSOR308110-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 2,6-Diazaspiro[3.3]heptane-2-carboxylate
Catalog # APOSOR308110-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :198.27 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1041026-70-3
Formula:C₁₀H₁₈N₂O₂
MDL Number:MFCD11226963