
1H-Pyrrolo[2,3-b]pyridin-4-amine
Catalog # APOSOR308038-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:133.15 g/mol
Melting Pt:122…123 °C
:GHS07
Storage Temperature:Ambient
CAS Number:74420-00-1
Formula:C₇H₇N₃
MDL Number:MFCD07778366