2-Chloro-3-(chloromethyl)-6,7-dimethylquinoline
Catalog # APOSOR307905-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.128 g/mol
Storage Temperature:Ambient
CAS Number:182052-67-1
Formula:C₁₂H₁₁Cl₂N
MDL Number:MFCD09787653