Order Entry
Italy
ContactUsLinkComponent
5-Boc-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Irritant
5-Boc-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Catalog # APOSOR307763-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Boc-1,4,6,7-tetrahydropyrazolo[4,3-c]pyridine
Catalog # APOSOR307763-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.28 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:230301-11-8
Formula:C₁₁H₁₇N₃O₂
MDL Number:MFCD08059268