Order Entry
Italy
ContactUsLinkComponent
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95+%
Irritant
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95+%
Catalog # APOSOR307073-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
cis-N-(tert-Butoxycarbonyl)-4-hydroxy-D-proline 95+%
Catalog # APOSOR307073-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.25 g/mol
Melting Pt:146…151 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:135042-12-5
Formula:C₁₀H₁₇NO₅
MDL Number:MFCD02094407