Order Entry
Italy
ContactUsLinkComponent
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%
Catalog # APOSOR305461-250MG
Supplier:  Apollo Scientific
CAS Number:  
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%
Catalog # APOSOR305461-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :191.19 g/mol
Storage Temperature:Ambient
CAS Number:1234616-14-8
Formula:C₉H₉N₃O₂
MDL Number:MFCD16658534