Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Methylsulphonyl)-3-pyridinamine
6-(Methylsulphonyl)-3-pyridinamine
6-(Methylsulphonyl)-3-pyridinamine
APOSOR303789-500MG
:
Apollo Scientific
:
6-(Methylsulphonyl)-3-pyridinamine
APOSOR303789-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
172.21 g/mol
:
175…177 °C
:
Ambient
:
187143-22-2
:
C₆H₈N₂O₂S
:
MFCD09941635