Order Entry
Italy
ContactUsLinkComponent
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine
Catalog # APOSOR302485-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine
Catalog # APOSOR302485-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :255.7200 g/mol
Storage Temperature:Ambient
CAS Number:16234-15-4
Formula:C₁₀H₁₀ClN₃OS
MDL Number:MFCD09909656