Order Entry
Italy
ContactUsLinkComponent
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine
Catalog # APOSOR302349-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-7-methylthieno[3,2-d]pyrimidine
Catalog # APOSOR302349-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.0900 g/mol
Storage Temperature:Ambient
CAS Number:35265-83-9
Formula:C₇H₄Cl₂N₂S
MDL Number:MFCD08061253