2-(1-Pyrazolyl)pyridine
Catalog # APOSOR302344-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:145.17 g/mol
Storage Temperature:Ambient
CAS Number:25700-11-2
Formula:C₈H₇N₃
MDL Number:MFCD09033534