

3-Cyanobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56542-67-7
:
Formula:C₇H₄ClNO₂S
MDL Number:MFCD00203480
Boiling Pt:114…116 °C/0.2mmHg
Melting Pt:47…53 °C
Flash Pt:>110 °C
UN:3261
ADR:8,II
:201.63 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together