Order Entry
Italy
ContactUsLinkComponent
tert-Butyl (4-formylpyridin-2-yl)carbamate
tert-Butyl (4-formylpyridin-2-yl)carbamate
Catalog # APOSOR302225-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl (4-formylpyridin-2-yl)carbamate
Catalog # APOSOR302225-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.24 g/mol
Storage Temperature:Ambient
CAS Number:304873-65-2
Formula:C₁₁H₁₄N₂O₃
MDL Number:MFCD09757496