3-Formyl-4-methoxypyridine
Catalog # APOSOR302197-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:137.14 g/mol
Storage Temperature:Ambient
CAS Number:82257-15-6
Formula:C₇H₇NO₂
MDL Number:MFCD04112505