Order Entry
Italy
ContactUsLinkComponent
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid 97%
Irritant
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid 97%
Catalog # APOSOR302114-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1H-Pyrrolo[3,2-c]pyridine-2-carboxylic acid 97%
Catalog # APOSOR302114-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.1500 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:800401-65-4
Formula:C₈H₆N₂O₂
MDL Number:MFCD09743299