Order Entry
Italy
ContactUsLinkComponent
6-Benzyl-2,4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine
Irritant
6-Benzyl-2,4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine
Catalog # APOSOR302097-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Benzyl-2,4-dichloro-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine
Catalog # APOSOR302097-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.19 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:778574-06-4
Formula:C₁₄H₁₃Cl₂N₃
MDL Number:MFCD08273934