Order Entry
Italy
ContactUsLinkComponent
1,3-Dichloro-2-iodobenzene 98%
Irritant
1,3-Dichloro-2-iodobenzene 98%
Catalog # APOSOR301367-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dichloro-2-iodobenzene 98%
Catalog # APOSOR301367-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :272.90 g/mol
Melting Pt:64…67 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:19230-28-5
Formula:C₆H₃Cl₂I
MDL Number:MFCD00060658