Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dihydro-2H-pyran-3(4H)-one
Dihydro-2H-pyran-3(4H)-one 95+%
Dihydro-2H-pyran-3(4H)-one 95+%
APOSOR301349-1G
:
Apollo Scientific
:
Dihydro-2H-pyran-3(4H)-one 95+%
APOSOR301349-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
100.1158 g/mol
:
GHS07
:
Refrigerator
:
23462-75-1
:
C₅H₈O₂
:
MFCD00182426