Order Entry
Italy
ContactUsLinkComponent
4-Chloro-6,7-dimethoxyquinoline
4-Chloro-6,7-dimethoxyquinoline
Catalog # APOSOR301095-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-6,7-dimethoxyquinoline
Catalog # APOSOR301095-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :223.6556 g/mol
Storage Temperature:Ambient
CAS Number:35654-56-9
Formula:C₁₁H₁₀ClNO₂
MDL Number:MFCD07778437