
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:201162-53-0
:MFCD11111771
Formula:C11H20N2O2
:212.29 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together