Order Entry
Italy
ContactUsLinkComponent
3-Formyl-2-methoxypyridine
3-Formyl-2-methoxypyridine
Catalog # APOSOR301072-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Formyl-2-methoxypyridine
Catalog # APOSOR301072-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :137.14 g/mol
Storage Temperature:Ambient
CAS Number:71255-09-9
Formula:C₇H₇NO₂
MDL Number:MFCD04115112