Order Entry
Italy
ContactUsLinkComponent
Methyl-3-(bromomethyl)benzoate
Corrosive
Methyl-3-(bromomethyl)benzoate
Catalog # APOSOR30105-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Methyl-3-(bromomethyl)benzoate
Catalog # APOSOR30105-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:43…46 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :229.07 g/mol
 :GHS05
CAS Number:1129-28-8
Formula:C₉H₉BrO₂
MDL Number:MFCD00051437