3-Methyl-2-(morpholin-4-yl)butanenitrile hydrochloride
Catalog # APOSOR300896-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.7 g/mol
Storage Temperature:Ambient
CAS Number:1672675-26-1
Formula:C₉H₁₇ClN₂O
MDL Number:MFCD28099771