
4-(2-Hydroxyethyl)benzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69395-13-7
:
Formula:C₉H₉NO
MDL Number:MFCD00052461
:147.18 g/mol
Boiling Pt:116…118 °C/0.5mmHg
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together