Order Entry
Italy
ContactUsLinkComponent
Ethyl-5-methyl-1H-pyrazole-4-carboxylate
Irritant
Ethyl-5-methyl-1H-pyrazole-4-carboxylate
Catalog # APOSOR300792-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-5-methyl-1H-pyrazole-4-carboxylate
Catalog # APOSOR300792-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.17 g/mol
Melting Pt:49…55 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:85290-78-4
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD08443874