
Ethyl pyrazole-3-carboxylate 99%
Catalog # APOSOR300747-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:140.14 g/mol
Melting Pt:159…161 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5932-27-4
Formula:C₆H₈N₂O₂
MDL Number:MFCD00159643