2-(4-Chlorophenyl)-2-morpholinoacetonitrile
Catalog # APOSOR300518-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Tipo di conf.:Bottle
- Conf:1 g
- Seq:3047
Specifications
About this item
:
:236.7 g/mol
Storage Temperature:Ambiente
CAS Number:33599-26-7
Formula:C₁₂H₁₃ClN₂O
MDL Number:MFCD00173106