Order Entry
Italy
ContactUsLinkComponent
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 95%
Irritant
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 95%
Catalog # APOSOR300503-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Hydroxy-1H-pyrazole-4-carboxylic acid ethyl ester 95%
Catalog # APOSOR300503-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:7251-53-8
Formula:C₆H₈N₂O₃
MDL Number:MFCD12131138