Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pagina principale
1-(4-Chlorophenyl)-2-thiourea
1-(4-Chlorophenyl)-2-thiourea
1-(4-Chlorophenyl)-2-thiourea
1-(4-Chlorophenyl)-2-thiourea
APOSOR29695-1G
:
Apollo Scientific
:
1-(4-Chlorophenyl)-2-thiourea
APOSOR29695-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
177…181 °C
:
Ambient
:
223-022-4
:
6.1,II
:
186.66 g/mol
:
2811
:
GHS06
:
3696-23-9
:
(C₆H₄Cl)NC(S)NH₂
:
MFCD00022168